Learn More
Honeywell™ North™ Lightning™ Series Safety Eyewear Replacement Lenses
Description
- Anti-scratch, anti-UV 99.9% and anti-static lenses
- Available in clear, smoke or mirrored tints
- Meets ANSI Z87.1-2010 and tested to meet the requirements of the CSA Z94.3 standard
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Replacement lens |