Learn More
Honeywell™ North™ Lightning™ T6500 Series Safety Eyewear
Description
- High base panoramic lens for a wide unobstructed field of view
- Frame made of new Elasto LT chemical resistant material
- New Soft-Touch comfort pads
- Self-adjusting temples that insure a secure yet comfortable fit
- The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
- Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
- Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
Specifications
Specifications
| Lens Coating | Anti-static, Scratch-resistant, UV-treated |
| Lens Tint | Smoke |
| Frame Color | Black |
| Temple Style | Self-adjusting |