Learn More
Honeywell™ North™ Lightning™ T6550 Series Safety Eyewear
Catalog No. 191300756
Change view
Click to view available options
Frame Color:
Black
Blue
Bronze
Silver
Lens Tint:
Amber
Mirror
Silver
Smoke
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-130-0756 Blue Amber
19-130-0752 Black Amber
19-130-0757 Blue Mirror
19-130-0755 Blue Smoke
19-130-0753 Black Mirror
19-130-0764 Bronze Amber
19-130-0765 Bronze Mirror
19-130-0763 Bronze Smoke
19-130-0758 Silver Silver
19-130-0761 Silver Mirror
19-130-0751 Black Smoke
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-130-0756 Supplier Honeywell™ Supplier No. T65505BLA
This item is not returnable. View return policy
Engineered for safety officers but designed for users
  • High base panoramic lens for a wide unobstructed field of view
  • Bowed temples designed to fit virtually anyone
  • New Soft-Touch comfort pads
  • Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
  • The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
  • Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-static, Scratch-resistant, UV-treated
Lens Tint Amber
Frame Color Blue
Temple Style Bowed w/soft-touch pads
Lens Material Polycarbonate