Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Powder-Free Latex Finger Cots

Catalog No. 19023276 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-023276 Small
19-023279 Medium
19-069930 Large
19-062392 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-023276 Supplier Honeywell™ North™ Supplier No. 115LWR/S
Add to Cart
Add to Cart
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Latex construction seals out moisture and debris, while also withstanding abrasions and punctures. Ideal for protecting fingertips and reducing finger prints when a full glove is unnecessary.

  • Latex construction seals out moisture and debris
  • 3 mil thickness is capable of withstanding punctures and abrasion
  • Uniform thickness ensures equal sensitivity in all critical areas
  • Very low extractables to reduce contamination from reactive ions
  • Finger cots are disposable to help reduce cross contamination
  • Provides an ideal level of protection when a full glove is unnecessary

Specifications

Applications Industrial
Color White
Disposable Single-use
Exterior Finish No Texture
Hand Ambidextrous
Material Latex
Powdered Powder Free
Product Type Finger Cot
Size Small
Thickness (English) 3 mil
For Use With (Application) Protecting Fingertips, Reducing Fingerprint Marks
Glove Cuff Style Rolled
Length (English) 2.8 in.
Length (Metric) 7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.