Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Nitri Task™ Nitrile Coated Nylon Gloves

Catalog No. 191500952
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0952 Large
19-150-0950 Small
19-150-0951 Medium
19-150-0954 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-150-0952 Supplier Honeywell™ Supplier No. NF13/9L
This item is not returnable. View return policy

Lightweight, 13-gauge gloves feature nitrile coating on the palm and fingertips for enhanced grip capabilities.

  • Lightweight, 13-gauge nylon
  • Nitrile coating on the palm and fingertips
  • Knit wrist prevents dirt and debris from entering the glove
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Gray, White
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Automotive Assembly, General Maintenance, Manufacturing, and Small Parts Handling
Glove Cuff Style Knit
Series NF13
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.