Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ NorthFlex Light Task™ Polyurethane Coated Nylon Gloves

Catalog No. 191500955
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0955 Small
19-150-0956 Medium
19-150-0957 Large
19-150-0958 X-Large
19-150-0959 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-150-0955 Supplier Honeywell™ North™ Supplier No. NF15/7S
This item is not returnable. View return policy

Polyurethane coated gloves are lightweight for optimum dexterity, while also providing a high level of mechanical protection.

  • Gloves combine dexterity with a high level of mechanical protection
  • 15 gauge nylon liner
  • Polyurethane coating on the palm
  • Launderable
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Automotive Assembly, Electronics, General Maintenance, Manufacturing, and Small Parts Handling
Glove Cuff Color Natural
Glove Cuff Style Knit
Series NF15
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.