Learn More
Honeywell™ North™ Polysafe and Polysafe Plus Wraparound Safety Glasses
Catalog No. 17985361
Change view
Click to view available options
Lens Tint:
Gray
No. per Pack:
72/Cs.
Catalog No. Lens Tint No. per Pack
17-985-361 Gray 72/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 17-985-361 Supplier Honeywell™ Supplier No. 11180032
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
One-piece construction provides UV absorption and impact protection from any angle
  • Sideshields comply with the OSHA Bloodborne Pathogens Standard, so spectacles are ideal for clinical use
  • Fit over many types of prescription eyewear
  • Polysafe Plus lenses are silicone-coated for scratch resistance and reduced glare
  • Polysafe Polycolors spectacles have the same clear lenses as Polysafe spectacles, but with colored temples
  • Approximate weight: 39g
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Gray
No. per Pack 72/Cs.