Learn More
Honeywell™ North™ Premier 611 Half-Mask Respirators for Paints and Pesticides
Description
- Complies with NIOSH 42 CFR 84 standards
- Half-mask design same as Series 6100 respirator
- Easy-to-clean silicone
- Dual-cartridge features a soft sealing flange and pliable nosepiece for maximum comfort
- Nonslip cradle suspension and floating yoke ensure a secure fit
- Additional room for protective eyewear
- P95 filter for paints and pesticides containing oil; the N95 filter is suitable for non-oil environments
Specifications
Specifications
| Material | Silicone |
| Product Type | Half-Mask Respirator |
| Size | Large |
| For Use With (Application) | Chemical, construction, food services, forestry, government, manufacturing, military, mining, municipal services, oil and gas, pharmaceutical, steel and metals, welding |
| For Use With (Equipment) | T Series Cartridge |
| Includes | Cartridges and Filter |
| Model | 313110 |