Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Protective Shell Inserts

Catalog No. 19166348 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Protective Shell Insert
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-166-348 Protective Shell Insert
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-166-348 Supplier Honeywell™ Supplier No. SC01H5
  • $188.50
    $150.65 / Pack of 6

    Save $37.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Protective Shell Insert
Color White
Material Polyethylene
For Use With (Application) Baseball cap, used by mechanics, muffler shops, golf course personnel, truck drivers, Oil change centers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.