Learn More
Honeywell™ North™ Protecto-Shield Replacement Visors
Tough faceshield protection with PresLock or Ratchet suspension adjustable headgear. Fits most commonly used visors.
Products 2
Catalog Number Mfr. No. Lens Material Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Material Lens Tint Price Quantity & Availability  
18-999-4540
View Documents
Honeywell™
11390046
Polycarbonate Dark Green N/A N/A N/A
18-999-4541
View Documents
Honeywell™
11390056
Aluminum-bound Acetate Medium Green N/A N/A N/A
Description
Description
  • Constructed of tough thermoplastic for superior heat protection
  • Designed with a chemical splash drip edge to prevent fluids from splashing down inside the visor
  • Easy sizing and tension adjustment for individual visor positioning
  • Visors easily attach with no snaps, posts or levers
  • Available with ratchet headband with tension knob that allows easy sizing and adjustment for individual visor positioning
  • Available with pinlock headband that provides adjustments similar to a baseball cap
  • Available in propionate, polycarbonate and acetate for a variety of environments
  • Popular industry sizes and thicknesses
  • Easy attachment without snaps, posts or levers; reinforced edge to resist cracking
  • Universal hole pattern allows the use of Sperian visors with other popular manufacturers' headgear
  • Aluminum bound acetate replacement visors form easily for a contoured, customized fit
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.