Learn More
Honeywell™ North™ Protecto-Shield Shaded Visors
Catalog No. 190881116 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Shade 4
Catalog No. Lens Tint
19-088-1116 Shade 4
1 options
This item is not returnable. View return policy
Catalog No. 19-088-1116 Supplier Honeywell™ Supplier No. 11390067
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tough faceshield protection with PresLock or Ratchet suspension adjustable headgear. Fits most commonly used visors.
  • Constructed of tough thermoplastic for superior heat protection
  • Designed with a chemical splash drip edge to prevent fluids from splashing down inside the visor
  • Easy sizing and tension adjustment for individual visor positioning
  • Visors easily attach with no snaps, posts or levers
  • Available with ratchet headband with tension knob that allows easy sizing and adjustment for individual visor positioning
  • Available with pinlock headband that provides adjustments similar to a baseball cap
  • Available in propionate, polycarbonate and acetate for a variety of environments
  • Popular industry sizes and thicknesses
  • Easy attachment without snaps, posts or levers and reinforced edge to resist cracking
  • Universal hole pattern allows the use of Sperian visors with other popular manufacturers' headgear
  • Aluminum bound acetate replacement visors form easily for a contoured, customized fit
Specifications
Lens Tint Shade 4
Product Type Replacement visor
Material Molded Propionate