Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Refresher Respirator Wipe Pads with Alcohol

Catalog No. 176234 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Respirator Wipes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-623-4 Respirator Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-4 Supplier Honeywell™ Supplier No. 7003H5
  • $105.08
    $78.65 / Pack of 100

    Save $26.43

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,050.81
    $786.65 / Case of 10 PK

    Save $264.16

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-moistened wipes provide a quick and easy way to clean respiratory equipment.

  • Removes perspiration and body oils from facepiece between scheduled cleanings
  • Contains 70% alcohol
  • Improve comfort, promote compliance
  • Wipe pad size, 5 x 7 in. (12.7 x 17.8cm)
  • Individually wrapped pads are sold 100 pads per box

Specifications

Product Type Respirator Wipes
Model 7003-H5
Solution Type Alcohol
For Use With (Application) Removal of perspiration and body oils from facepiece between scheduled cleanings
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.