Learn More
Honeywell™ North™ Replacement Hoods for Airline Respirators
Catalog No. 19034691C
Change view
Click to view available options
Replacement Hood
Catalog No.
19-034-691C Replacement Hood
1 options
This item is not returnable. View return policy
Catalog No. 19-034-691C Supplier Honeywell™ Supplier No. 85302TA
This item is not returnable. View return policy
Available in packs of 10.
TRUSTED_SUSTAINABILITY
Specifications
Material Tyvek
Product Type Respirator Hood
For Use With (Application) Excellent protection against airborne contaminants in a variety of applications including spray painting, abatement, pharmaceutical manufacturing and chemical handling
For Use With (Equipment) Type c continuous flow airline respirator
Model 85302TA-H5
Quantity 1 Ea.