Learn More
Honeywell™ North™ Series 7900 Emergency Escape Mouthbit Respirator
Catalog No. 19816212 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Acid Emergency Escape Mouthpiece Respirator
Catalog No.
19-816-212 Acid Emergency Escape Mouthpiece Respirator
1 options
This item is not returnable. View return policy
Catalog No. 19-816-212 Supplier Honeywell™ Supplier No. 7902
  • $96.40
    $54.65 / Each
    Save $41.75
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $838.00
    $546.65 / Case of 10 EA
    Save $291.35
    35% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 7900 Series mouthbit respirators offer a quick and economical method to escape non-IDLH emergency situations. Use the respirator as a visitor escape device or issue individual respirators to employees as a safety precaution against a potential chemical leak
  • Provides escape from acid gas or ammonia
  • Convenient neckstrap or beltclip
  • Lightweight and disposable
Model 7902 is designed for “escape only” from atmospheres containing chlorine, chlorine dioxide, sulfur dioxide, and hydrogen sulfide. Use Model 7904 for environments where ammonia is present.
Low breathing resistance increases wearer acceptance and reduces fatigue. Exhalation valve prevents inward contamination leakage; inhalation valve keeps exhaled breath and spittle from contacting chemical absorbent area. Break-away neck strap reduces risk of injury should respirator become caught in machinery. Large stainless-steel belt clip ensures easy attachment to belt or pants.
Specifications
Certifications/Compliance NIOSH
Product Type Emergency Escape Mouthpiece Respirator
For Use With (Application) Emergency use at pulp and paper mills, water treatment plants, pools and spas, chemical plants and other facilities
Includes Single cartridge, mouthpiece, neck strap and nose clip
Size Universal