Learn More
Honeywell™ North™ Silicone Full Facepiece Accessories, Eyeglass Frame
Catalog No. 19816181 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Eyeglass Frame
Catalog No.
19-816-181 Eyeglass Frame
1 options
This item is not returnable. View return policy
Catalog No. 19-816-181 Supplier Honeywell™ Supplier No. 80100H5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accessory for use with 5400 and 7600 series full facepiece.
  • Eyeglass frame accessory
  • For use with 5400 and 7600 series full facepieces
  • Honeywell model 80100-H5
Specifications
Certifications/Compliance ANSI Z136.1
Material Plastic
Product Type Eyeglass Frame
For Use With (Application) Manufacturing, medical
For Use With (Equipment) 5400 and 7600 series full facepiece
Includes Cleaning cloth or a micro-fiber cloth bag (which doubles as a cleaning cloth), a hard-case for storage and a safety hang cord to carry the eyewear around your neck
Model 80100-H5