Learn More
Honeywell™ North™ Vapor Safety Glasses
Catalog No. 191300844 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Lens Coating:
Hardcoat
Catalog No. Lens Tint Lens Coating
19-130-0844 Clear Hardcoat
1 options
This item is not returnable. View return policy
Catalog No. 19-130-0844 Supplier Honeywell™ Supplier No. 11150900
  • $17.50
    $11.65 / Each
    Save $5.85
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $174.71
    $116.65 / Pack of 10 EA
    Save $58.06
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Capture sports styling in a painted, metallic blue frame
Capture sports styling in a painted, metallic blue frame
•    9–base wrap-around dual lenses and sport temples offer a comfortable secure fit
•    Soft nose bridge prevents slipping
•    Durable polycarbonate lens feature scratch-resistant or Fog-Ban™ antifog coating
•    Adjustable breakaway Flexicord™ included
Compliance
Meet ANSI Z87+ and CSA Z94.3 safety standards
Specifications
Lens Coating Hardcoat
Certifications/Compliance Meet ANSI Z87+ and CSA Z94.3 safety standards
Lens Tint Clear
Includes Break-Away Flexicord
Temple Style Sport
Lens Material Polycarbonate