Learn More
Honeywell™ North™ XV100 Series Safety Glasses
Catalog No. 17377452
Change view
Click to view available options
Frame Color:
Clear Frosted
Frost Gray
Lens Coating:
Hardcoat
Scratch-resistant
Lens Tint:
Clear
Gray
Silver Mirror, Indoor/Outdoor
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
17-377-452 Clear Frosted Hardcoat Clear
17-377-455 Clear Frosted - Clear
17-377-453 Frost Gray - Gray
17-377-454 Clear Frosted Scratch-resistant Silver Mirror, Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-377-452 Supplier Honeywell™ Supplier No. XV100
This item is not returnable. View return policy
Excellent peripheral protection and optical clarity
  • 10-base, wraparound uni-lens for great peripheral vision and protection
  • Molded, universal nosepiece for easy fit
  • Lightweight design and thin, shaped temples for all-day comfort
Compliance
Meet ANSI™ Z87.1-2003 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Hardcoat
Certifications/Compliance Meet ANSI Z87.1-2003 standards.
Lens Tint Clear
Frame Color Clear Frosted
No. per Case 200/Cs.
Temple Style Curved