Learn More
Honeywell North™ North Zone Ratchet Cap Style Hard Hat
Catalog No. 17378711 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Black
Gray
Hot Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-378-711 Hot Pink
17-378-707 Black
17-378-705 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-378-711 Supplier Honeywell North™ Supplier No. N10R200000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contemporary shell design with four large areas for custom logo imprinting or other personalization options
  • 3-level height adjustment (low, medium, high) for greater stability, balance and retention
  • Removable
    • Replaceable rear comfort cradle for improved fit and comfort
  • Easy-grip ratchet for smooth and precise headband adjustment
    • Even when wearing gloves
  • Ergonomic headband for adjustment to 3 different sizes (small, medium/large and X-large)
  • Moisture-wicking, breathable sweatband
    • Easily removeable, washable and replaceable
  • Short brim and low-profile design for increased field of vision and user mobility
  • Slots for cap-mounted earmuffs, face shields and other PPE accessories
Specifications
Certifications/Compliance ANSI/ISEA Z89.1 2014: Type I, Class C, G, and E, CSA Z94.1 2015: Type I, Class C, G, and E
Color Hot Pink
Product Type Short Brim Hard Hat
Size Universal
Ventilated Not Ventilated
Head Style Short Brim
Width (Metric) 21.84 cm
Width (English) 8.6 in.