Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NorthernLights Streptavidin NL493
SDP

Catalog No. NL997 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL997 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL997 Supplier R&D Systems™ Supplier No. NL997

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry, Immunohistochemistry, Immunocytochemistry

  • Phosphate-buffered saline (PBS) containing <0.1% v/v sodium azide
  • Absorption macimum 493nm; Emission maximum 514nm
  • Provided in liquid format

Applications:

Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Quantity 0.5 mL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.