Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noscapine hydrochloride hydrate, MilliporeSigma™ Supelco™

Catalog No. 111015894 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5894 25 g
11-101-5895 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5894 Supplier MilliporeSigma Supelco Supplier No. N900725G
Add to Cart
Add to Cart
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 912-60-7
Physical Form Neat
Linear Formula C22H23 NO7 · HCl · xH2 O
MDL Number MFCD00149223
Synonym Narcotine hydrochloride
Type Analytical Standard
Quantity 25 g
Formula Weight 449.88 (anhydrous basis)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.