Learn More
Cell Signaling Technology NOTCH ISOFORM ANTIBODY SAMPLER
SDP
Supplier:  Cell Signaling Technology 3640T
Encompass
The Notch Isoform Antibody Sampler Kit provides an economical means to investigate Notch Signaling. The kit contains primary and secondary antibodies to perform two western mini-blots with each antibody.
Catalog No. NC2596596
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.