Learn More
Enzo Life Sciences Notch2 Recombinant monoclonal antibody (B6) (Mouse IgG1 kappa) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6490100
Encompass_Preferred
Clone: B6. Isotype: IgG1κ. Immunogen: NRR (negative regulatory region) of mouse Notch2. Species reactivity: Human, Mouse. Specificity: Recognizes both mouse mNRR2 and human hNRR2, but not NRR1. Applications: ELISA, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3173
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.