Learn More
Invitrogen™ NOTCH2NL Polyclonal Antibody
Description
NOTCH2NL Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | NOTCH2NL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | NOTCH2NL |
| Gene Accession No. | Q7Z3S9 |
| Gene Alias | N2N; notch 2 N-terminal like; Notch homolog 2 N-terminal like protein; notch homolog 2 N-terminal-like protein; Notch homolog 2 N-terminal-like protein A; Notch homolog 2 N-terminal-like protein C; NOTCH2NL; NOTCH2NLA; NOTCH2NLC |
| Gene Symbols | NOTCH2NL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.