Learn More
Invitrogen™ NOTCH3 Monoclonal Antibody (MHN3-21), APC, eBioscience™

Description
Description: This MNH3-21 monoclonal antibody reacts with the extracellular domain of human Notch3, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Notch3 expression has been demonstrated on some thymocyte subsets, including CD4-CD8- and CD8SP cells. This Notch receptor is also expressed on vascular smooth muscle and cells of the central nervous system. In addition to its role in stem cell hematopoiesis, Notch3 plays a pivotal role in T cell lineage commitment and thymocyte development. Moreover, Notch3 is overexpressed in human T-cell acute lymphoblastic leukemias (T-ALL) and other cancers. Finally, mutation of Notch3 has been linked to cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL), a CNS degenerative disorder. Applications Reported: This MHN3-21 antibody has been re...
Specifications
Specifications
| Antigen | NOTCH3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MHN3-21 |
| Concentration | 5 μL/Test |
| Conjugate | APC |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Notch3 |
| Gene Accession No. | Q9UM47 |
| Gene Alias | AW229011; CADASIL; CADASIL1; CASIL; hpbk; IMF2; LMNS; N3; Neurogenic locus notch homolog protein 3; notch 3; Notch 3 extracellular truncation; Notch 3 intracellular domain; Notch gene homolog 3; Notch homolog 3; NOTCH3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.