Learn More
Simport™ Scientific Notched Silicone Sealing Mats for Deep Well Plates, 96 Square
Shop All Simport Scientific ProductsDescription
- Effectively seals deep well plates
- Easy to open and close
- Prevents the volatilization of a variety of solvents
- Chemical and heat resistant
- All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
- Resistant to DMSO and biological buffers
- Free of RNase, DNase, Pyrogen, and DNA
Specifications
Specifications
| Color | Natural |
| Solvent Or Alcohol Resistant | Alcohol Resistant, Solvent Resistant |
| Material | Silicone |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| Pre-slit | Yes |
| Certified RNase/DNase-free | Yes |
| Pierceable | Yes |
| Temperature Range (Metric) | -80°C to +80°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.