Learn More
Invitrogen™ NOTUM Polyclonal Antibody

Description
A suggested positive control for this product is HepG2 cell lysate.
Specifications
Specifications
| Antigen | NOTUM |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | NOTUM |
| Gene Accession No. | Q6P988, Q8R116 |
| Gene Alias | [Wnt protein] O-palmitoleoyl-L-serine hydrolase; 5730593N15Rik; hNOTUM; NOTUM; notum pectinacetylesterase homolog; notum pectinacetylesterase homolog (Drosophila); NOTUM, palmitoleoyl-protein carboxylesterase; OK/SW-CL.30; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog; RGD1307119 |
| Gene Symbols | NOTUM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.