Learn More
Drg International Inc Nov Elisa
Supplier:  Drg International Inc EIA4836
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of NOV in serum, plasma and urine. This product is for Research Use Only. The NOV ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal (mouse) antibody directed towards a unique antigenic site of the NOV molecule. An aliquot of patient sample containing endogenous NOV is incubated in the coated well together with the assay buffer. After washing, a biotinylated anti-NOV antibody (enzyme conjugate) is added. After incubation, the unbound conjugate is washed off and Streptavidin conjugated horseradish peroxidase (Enzyme complex) is added. The amound of bound peroxidase is proportional to the concentration of NOV in the patient sample.
Catalog No. 50-194-7619
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.