Learn More
Description
- Polycationic liposomal formulation
- Ideal for mammalian protein production
- Derived from non-animal sources
- Minimal cellular toxicity
- Sterile ready to use solution
- Optimized for transient transfections of HEK 293 suspension cultures
- Compatible with both serum-containing serum-free media
- Eliminates need for media changes
- Packaging: Glass Bottle
Specifications
Specifications
| Content And Storage | 2°C to -8°C |
| Quantity | 5 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.