Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ 2xYT Broth

Catalog No. 717554 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-755-4 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-755-4 Supplier MilliporeSigma™ Supplier No. 717554
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the enrichment of E. coli. MilliporeSigma™; Novagen™ 2xYT Broth is a powdered medium.

  • Composition of the 2xYT Broth per liter is 16g tryptone, 10g yeast extract, and 5g sodium chloride
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Microbiology Media
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.