Learn More
MilliporeSigma™ Novagen™ 4X SDS Sample Buffer
Catalog No. 706073 Shop All MilliporeSigma Products
Change view
Click to view available options
Concentration:
4X
Catalog No. Concentration
70-607-3 4X
1 options
Catalog No. 70-607-3 Supplier MilliporeSigma™ Supplier No. 706073
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for SDS-PAGE analysis of proteins.
EMD Millipore Novagen™ 4X SDS Sample Buffer is stard formulation
  • Includes DTT for complete denaturation of disulfide bonds
  • Can be used at 2X for most applications
TRUSTED_SUSTAINABILITY
Specifications
Concentration 4X
Product Type DNA Sample Loading Buffer