Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ 5% Alkali - Soluble Casein

Catalog No. 709553 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
225 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-955-3 225 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-955-3 Supplier MilliporeSigma™ Supplier No. 709553

missing translation for 'validMainframeMsg'

Used as blocking reagent for Western blot applications.

EMD Millipore Novagen™ 5% Alkali - Soluble Casein used in colorimetric chemiluminescent detection protocols.

  • Minimizes non-specific background
  • High coating properties, detection sensitivity
  • Concentration optimized for maximal signal noise ratios
  • Recommended working concentration range: 0.25-1%
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Quantity 225 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.