Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ 6X Loading Buffer

Catalog No. 691803 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-180-3 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-180-3 Supplier MilliporeSigma™ Supplier No. 691803
Add to Cart
Add to Cart

Intended for gel electrophoresis of DNA fragments or PCR products.

EMD Millipore Novagen™ 6X Loading Buffer contains Orange G, Xylene Cyanole FF glycerol.

  • Applications: Gel electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Description 6X Loading Buffer
Concentration 6X
Quantity 5 mL
Storage Requirements 2° to 8°C
Product Type DNA Sample Loading Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.