Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ BL21 Competent Cells

Catalog No. 694493 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-449-3 0.4 mL
69-449-4 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-449-3 Supplier MilliporeSigma™ Supplier No. 694493
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BL21 host strain is the most widely used host background and has the advantage of being deficient in both lon and ompT proteases.

BL21 is the most widely used host background and has the advantage of being deficient in both
TRUSTED_SUSTAINABILITY

Specifications

Contains F' Episome Lacks F' Episome
Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage ≤-70°C
Blue/White Screening No
Quantity 0.4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.