Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ λDE3 plus pLysS and pLysE Lysogenization Kit

Catalog No. 697253 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-725-3 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-725-3 Supplier MilliporeSigma™ Supplier No. 697253
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for site-specific integration of λDE3 prophage into E. coli host cell chromosome. MilliporeSigma™; Novagen™ λDE3 plus pLysS and pLysE Lysogenization Kit is designed such that the lysogenized host can be used to express target genes cloned in pET vectors.

  • Used in genomic analysis applications
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Genomic Analysis
Quantity 10 Reactions
Product Type λDE3 plus pLysS and pLysE Lysogenization Kit
Content And Storage Store at ≤70°C. Avoid freeze/thaw. Ok to freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.