Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ DNase I, RNase free

Catalog No. 691823 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-182-3 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-182-3 Supplier MilliporeSigma™ Supplier No. 691823
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For critical maintenance of RNA integrity applications

  • Digests either single- or double-stranded DNA, producing mixture of mono- and oligonucleotides
  • Selectively degrades DNA in presence of RNA
  • Can be used to remove DNA template following in vitro transcription reactions
  • Used in DNase footprinting and nick translation
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 1000 U
Product Type DNase I (RNase-free)

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.