Learn More
Description
EMD Millipore Novagen™ GeneJuice™, Transfection Reagent is non-lipid based chemical transfection reagent.
- Efficient DNA transfer for both stable transient transfections
- Minimal cellular toxicity
- Compatibility with both serum-containing serum-free media
- Simple protocol
- Suitable for high-throughput transfection in multi-well plate format
- CAS: Ethanol: 64-17-5
- Packaging: Glass Bottle
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Quantity | 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.