Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ KOD DNA Polymerase
GSA_VA

Catalog No. 710853 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-085-3 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-085-3 Supplier MilliporeSigma™ Supplier No. 710853
Add to Cart
Add to Cart

For PCR amplification of DNA templates, general cloning, cDNA amplification applications

  • Higher fidelity than Pfu DNA polymerase that helps for cloning
  • Greater yield: Extension speed is 2X faster than Taq DNA polymerase and 5X faster than Pfu DNA polymerase
  • Higher processivity: Sequential nucleotide polymerization is 10- to 15-fold greater than Pfu and Tli DNA polymerases
  • Amplifies plasmid and lambda DNA templates up to 6 kbp
  • Amplifies genomic DNA templates up to 2 kbp
  • No truncated amplification products

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2.5 U/μL
Purity > 90% homogeneous by SDS-PAGE
Content And Storage -20°C
Source Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli
Quantity 250 U
Product Type KOD DNA Polymerase

For Research use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.