Learn More
MilliporeSigma™ Novagen™ KOD Hot Start Master Mix
GSA_VA
Catalog No. 718423 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 rxn
500 rxn
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-842-3 100 rxn
71-842-4 500 rxn
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-842-3 Supplier MilliporeSigma™ Supplier No. 718423
Add to Cart
Add to Cart
For convenient high fidelity PCR
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA template up to 21 kbp
  • Eliminates mispriming and primer-dimer formation
  • Convenient ambient-temperature setup compatible with automation
  • KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Product Type KOD Hot Start Master Mix
Quantity 100 rxn