Learn More
MilliporeSigma™ Novagen™ KOD Hot Start Master Mix
Catalog No. 718424 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 rxn
500 rxn
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-842-4 500 rxn
71-842-3 100 rxn
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-842-4 Supplier MilliporeSigma™ Supplier No. 718424
Add to Cart
Add to Cart
For convenient high fidelity PCR
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA template up to 21 kbp
  • Eliminates mispriming and primer-dimer formation
  • Convenient ambient-temperature setup compatible with automation
  • KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Product Type KOD Hot Start Master Mix
Quantity 500 rxn