Learn More
Description
- Prevents clumping of medium and inhalation of airborne powder
- Supplied in several formats
- EasyPak is aluminum foil pouch containing 37g granulated medium sufficient for 1L culture
- Add EasyPak contents to 1L water, microwave to dissolve agar and sterilize
- Supplied in 500g and 5kg bottles
- Composition of LB Agar MILLER per liter is 5g yeast extract, 10g peptone from casein, 10g sodium chloride, and 12g agar-agar
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Product Type | Agar |
| Quantity | 25 Easy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.