Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ NovaQUANT™ Coli qPCR Kit

Catalog No. 726331KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
726331KIT 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 726331KIT Supplier MilliporeSigma™ Supplier No. 726331KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For accurate detection of trace amounts of bacterial DNA, typically in low femtogram range

  • Stable quantitative reat-time PCR assay for E.coli genome stuides
  • Sensitively and linear detection of E.coli DNA from virtually any prepared sample
  • Eliminate or standardize for downstream process
  • Quality control extracted and processed nucleic acids to avoid erroneous sequence amplification or cloning
  • Qualify samples for NGS and downstream libraries to ensure high quality data output
  • Evaluates E.coli expressed recombinant proteins
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 200μL NovaQUANT Coli qPCR Primer Mix (2μM), 100 ng NovaQUANT Coli Control DNA (10ng/μL)
Description Coli qPCR Kit
Format Kit
Technique qPCR
Kit Type QPCR DNA Detection Kit
Quantity 1 kit
Reverse Transcriptase Not Included
Shipping Condition Dry Ice
Species E. coli
For Use With (Application) Real Time PCR (qPCR)
Final Product Type Bacterial DNA
Sample Type Extracted and Processed Nucleic Acids
Starting Material E. coli DNA
Volume 200 μL
Includes 200 μL NovaQUANT Coli qPCR Primer Mix (2 μM), 100 ng NovaQUANT Coli Control DNA (10 ng/μL)
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.