Learn More
MilliporeSigma™ Novagen™ NovaQUANT™ Mouse Oxidative Stress Kit
Catalog No. 726281KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Mouse Oxidative Stress Kit
Catalog No.
726281KIT Mouse Oxidative Stress Kit
1 options
Catalog No. 726281KIT Supplier MilliporeSigma™ Supplier No. 726281KIT
Designed to quantify relative mRNA levels of 21 transcriptionally-regulated analytes associated with oxidative stress responses and three house-keeping control genes used as controls
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2 each NovaQUANT Mouse Oxidative Stress qPCR Kit, 100 ng LM cDNA (10ng/μL); 100 ng Mouse Oxidative Stress qPCR Primer Plate
Description Mouse Oxidative Stress Kit
Quantity 1 kit