Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ NovaTaq™ DNA Polymerase

Catalog No. 710035 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 U
2500 U
500 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-003-5 2500 U
71-003-3 100 U
71-003-4 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-003-5 Supplier MilliporeSigma™ Supplier No. 710035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For dependable PCR amplification

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Purity > 95% homogeneous by SDS-PAGE
Content And Storage -20°C
Source Recombinant Thermus aquaticus DNA polymerase expressed in E. Coli
Quantity 2500 U
Product Type DNA Polymerase

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.