Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ NovaTaq™ Hot Start DNA Polymerase

Catalog No. 710913 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1250 U
250 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-091-3 250 U
71-091-4 1250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-091-3 Supplier MilliporeSigma™ Supplier No. 710913
Add to Cart
Add to Cart

For quantitative and high-throughput PCR applications

  • Heat-activatable modified form of recombinant Taq DNA polymerase
  • Higher PCR specificity and yield
  • Improved low-copy target amplification
  • Ambient temperature setup compatible with automation
  • Target amplification of up to 5 kbp
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage -20°C
Source Recombinant Thermus aquaticus DNA polymerase expressed in E. coli
Quantity 250 U
Product Type Hot Start DNA Polymerase

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.