Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ NovaTaq™ Hot Start DNA Polymerase

Catalog No. p-5538853 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1250 U
250 U

For quantitative and high-throughput PCR applications

  • Heat-activatable modified form of recombinant Taq DNA polymerase
  • Higher PCR specificity and yield
  • Improved low-copy target amplification
  • Ambient temperature setup compatible with automation
  • Target amplification of up to 5 kbp
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage -20°C
Source Recombinant Thermus aquaticus DNA polymerase expressed in E. coli
Quantity 250 U
Product Type Hot Start DNA Polymerase

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.