Learn More
MilliporeSigma™ Novagen™ NovaTaq™ Hot Start Master Mix Kits
Catalog No. 716763 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1000 rxn
200 rxn
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-676-3 200 rxn
71-676-4 1000 rxn
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-676-3 Supplier MilliporeSigma™ Supplier No. 716763
Only null left
Add to Cart
Add to Cart
Premixed 2X hot start PCR components for convenience and reproducibility
  • Simplifies set-up for PCR and offers time savings, consistency, and minimal risk of contamination
  • Provide sufficient components for 200 or 1000 standard 50μL amplification reactions
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Ready-to-use 2X mixture of NovaTaq Hot Start DNA Polymerase, ultrapure deoxynucleotides, and reaction buffer with MgCl2, PCR Grade Water, MgCl2 for optimizing Mg2+ concentrations
Product Type Hot start master mix kit
Quantity 200 rxn