Learn More
MilliporeSigma™ Novagen™ Oligo(dT) primer
Catalog No. 698963 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
69-896-3 20 μg
1 options
Catalog No. 69-896-3 Supplier MilliporeSigma™ Supplier No. 698963
Add to Cart
Add to Cart
Used for first-strand synthesis of cDNAs, using poly-adenylated mRNA as template
  • Can be used in the following systems:
  • OrientExpress™ Oligo(dT) cDNA Library Construction System
  • OrientExpress™ Oligo(dT) cDNA Synthesis Kit
  • T7Select™ 1-1 OrientExpress Oligo(dT) cDNA Cloning System
  • T7Select™ 10-3 OrientExpress Oligo(dT) cDNA Cloning System
TRUSTED_SUSTAINABILITY
Specifications
Product Type Oligo(dT) primer
Content And Storage -20°C
Quantity 20 μg