Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ One Step RT-PCR Master Mix Kit

Catalog No. 719783 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 rxn
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-978-3 50 rxn
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-978-3 Supplier MilliporeSigma™ Supplier No. 719783
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For gene expression studies from tissues and cells

  • Robust one-step, one-enzyme master mix system for easy reaction assembly
  • Eliminates risk of cross contamination associated with two-step RT-PCR protocols
  • High-temperature (60° C) for reverse transcription enhances read-through of RNA secondary structure
  • Optimized buffer conditions and antibody-mediated hot start for increased sensitivity
  • Rapid enzyme activation step (30 s) avoids damage of template RNA

Components:

  • 2 x 625μL 2X One Step RT-PCR Master Mix
  • 1 x 200μL 50 mM Mn(OAc)2
  • 1 x 1.1mL RNase Free Water
  • 1 x 50μL Primer F (10 pmol/μL, 5'-TCC ACC ACC CTG TTG CTG TA-3')
  • 1 x 50μL Primer R (10 pmol/μL, 5'-ACC ACA GTC CAT GCC ATC AC-3')
  • 1 x 50μL Positive Control RNA (5 x 108 copies/mL)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type One Step RT-PCR Master master mix kit
Quantity 50 rxn

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.