Learn More
MilliporeSigma™ Novagen™ Origami™ 2(DE3) Singles™ Competent Cells
Catalog No. 714084 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
11 Reactions
22 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-408-4 22 Reactions
71-408-3 11 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-408-4 Supplier MilliporeSigma™ Supplier No. 714084
Add to Cart
Add to Cart
Origami 2 strains have mutations in glutathione reducatase (gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation.
Origami™ 2 host strains are K-12 derivatives that have mutations in both the thioredoxin reductase (trxB) and glutathione reductase (gor) genes, which greatly enhance disulfide bond formation in the E. coli cytoplasm. The Origami 2 strains are kanamycin sensitive; like the original Origami strains, the gor mutation is still selected for by tetracycline. To reduce the possibility of disulfide bond formation between molecules, strains containing mutations in trxB and gor are recommended only for the expression of proteins that require disulfide bond formation for proper folding. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY
Specifications
Efficiency Guaranteed efficiency: 2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 22 Reactions