Learn More
MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction NMR Medium-15N
Shop All MilliporeSigma ProductsDescription
- Autoinduction method is based on the differential metabolism of carbon sources to promote growth to high cell density
- Automatically induce protein expression under control of lac promoters, eliminating the need to monitor cell densities or add inducer (IPTG)
- Includes four concentrated sterile solutions:
- OnEx NMR Solution 1 is a blend of carbon sources optimized for tightly regulated uninduced growth to high density, followed by autoinduction
- OnEx NMR Solution 2 is a buffer and nitrogen blend that mediates metabolic acid production and provides additional nitrogen for increased protein synthesis
- OnEx Solution 3 provides magnesium for maximal cell density
- OnEx Solution 4 provides trace metals to minimize growth limitations associated with mineral deficiencies and satisfy the requirements of metal-containing target proteins, even at high expression levels
Specifications
Specifications
| Quantity | 1 L Kit |
| Content And Storage | Multiple storage requirements. Ok to freeze. |
| Product Type | NMR Medium-15N |